(±)-Epichlorhydrin, GPR RECTAPUR®
25 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Epichlorhydrin, 1-Chlor-2,3-epoxypropan, (±)-1-Chlor-2,3-epoxypropan, 3-Chlor-1,2-epoxypropan, (±)-3-Chlor-1,2-epoxypropan
- CAS Number:106-89-8
- Formula:C₃H₅ClO
- MDL Number:MFCD00005132
- Molecular Weight:92.52 g/mol
- Merck Index:13,03642
- Boiling Pt:117,9 °C (1013 hPa)
- Melting Pt:–57,2 °C
- Density:0,9985 g/cm³ (20 °C)
- Flash Pt:28 °C
- Storage Temperature:Stue temperatur
- EINECS:203-439-8
- UN:2023
- ADR:6.1 (3),II
Documents
Certificates
Search by Lot NumberAvantor Anhydrous Solvents.pdf VWR Chemical Substances for Organic Synthesis VWR Chemicals Big Packs and Bulks VWR Chemicals for NMR and UV VIS Spectroscopy VWR® Green Solvents for Sustainable Chemistry.pdf
Product Literature
spec_VWRC87074.260 spec_VWRC87074.290 spec_VWRC87074.460
Product specification
Specifications
Specification Test Results
- Appearance :Clear liquid
- Assay (GC) :≥99.0 %
- Density (d 20 °C/ 4 °C) :1.178 - 1.182
- Identity (IR) :passes test
Specifications
- Pakn:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



