Mobile tables for centrifuges
Product Details & Documents
Free up your bench.
- For centrifuges 5804/5804 R, 5810/5810 R, 5910 R and 5920 R
Use the mobile tables if there is not enough space on the regular lab bench, or to lower the operating height of your centrifuge for a more ergonomic work environment. We can also transport and use the centrifuge in other labs. Whether choose the low design model for under bench storage of centrifuge or the high design model for ergonomic loading and unloading of rotors and samples, both mobile tables have a very sturdy design for safe centrifugation and are equipped with heavy-duty transport wheels and brakes, these follow the same extremely high standards in safety and reliability.
Specifikationer
- Beskrivelse:Mobile table for ergonomic loading and unloading (high design), fits in any empty floor space
- Seq:02
- B×D×H:749×701×584 mm
- Cat. No.:EPPE5921000.012
- Miljøcertificering:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Lav fremstillingspåvirkning