Orders LinkContactUsLinkComponent
Nailsaver scouring sponges
Nailsaver scouring sponges
rød/hvid, 133×70 mm
Katalog # MMMASCH106
Leverandør:   3M
Nailsaver scouring sponges
Katalog # MMMASCH106
Leverandør:  3M
Nailsaver scouring sponges
Nailsaver scouring sponges
rød/hvid, 133×70 mm
Price
128,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
3M™ Nailsaver scouring sponges are cleaning sponges made of a textured synthetic fibre white web containing very soft abrasive particles, laminated with an adhesive onto a grooved synthetic colour-coded foam holder.
  • Easy positioning four colours of foams) to comply with HACCP requirements
  • Good cleaning performance for light-duty cleaning tasks
  • Soft and low scratching power on most surfaces
  • Easy to handle and finger protection thanks to the shape of the foam
  • Very long lasting
For easy cleaning with low scratching power on most surfaces, use 3M™ Nailsaver scouring sponges.
These scouring sponges offer good cleaning performance for light-duty cleaning tasks with soft and low scratching power on most surfaces. They’re very long lasting and easy to handle thanks to the shape of the foam.
Specifikationer
  • Farve:
    rød/hvid
  • L×B:
    133×70 mm
  • Seq:
    03