PTFE tapes, ResTape
ResTape, compatible with a broad range of gases and liquids, gul
Katalog # REST22486
Leverandør: Restek
Gå til:
Product Details & Documents
ResTape PTFE tape is for use with threaded connections in a wide range of plumbing materials.
- Each roll is 12,7 mm × 6,6 m
Tapes have a maximum effective temperature of 260 °C.
Specifikationer
- Farve:gul
- Beskrivelse:ResTape, compatible with a broad range of gases and liquids
- Seq:02