Orders LinkContactUsLinkComponent
SafeKey lockout padlocks, nylon
SafeKey lockout padlocks, nylon
SafeKey padlock, 113,4 g, blå, 38×19×84 mm
Katalog # BRDY150251
Leverandør:   Brady
SafeKey lockout padlocks, nylon
Katalog # BRDY150251
Leverandør:  Brady
SafeKey lockout padlocks, nylon
SafeKey lockout padlocks, nylon
SafeKey padlock, 113,4 g, blå, 38×19×84 mm
Price
195,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These SafeKey nylon padlocks are the ultimate locks for all types of safety and security needs.
  • Lightweight with a ribbed design for easy gripping
  • Ideal for electrical applications and enclosures
  • Key releases only upon lock closure
  • Preferred for most indoor lockout applications
  • Engraving options on front, back, left side, right side, and/or key
  • Key retaining (Key won’t release until the padlock shackle is securely closed)
  • Non-conductive
The new design and innovative keying mechanism provides more unique key possibilities than a conventional standard safety padlock, without sacrificing a secure lockout. The unique lock mechanism is insulated and non-conductive to protect workers from shocks when key is inserted. The lock mechanism has six precision steel tumbler blades for a high number of key differences and tamper resistance.
Locks are lightweight and resist impact, temperature, chemicals, and corrosion. Their non-conductive, non-sparking body features a ribbed design for easier gripping and handling. The colour-matched key and lock body allows a fast identification in the application.
Ordering information: Custom keying, engraving, charting, Master Key, and Grand Master Key options are available.
Delivery information: One key is supplied with each lock.
Specifikationer
  • Farve:
    blå
  • Beskrivelse:
    SafeKey padlock, 113,4 g
  • Seq:
    04
  • B×D×H:
    38×19×84 mm