PC certified clean validation bottles, Nalgene®
20 ml, PC certified clean validation bottles
Katalog # 216-2288
Leverandør: Thermo Fisher Scientific
PC certified clean validation bottles, Nalgene®
Katalog # 216-2288
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
- Polycarbonate construction
- Sterile
- Clear finish
- Certified clean
These bottles can reduce the risk of contamination.
: Validated to USP 788 and <0,5 EU/ml endotoxin.
Specifikationer
- Kapacitet:20 ml
- Beskrivelse:PC certified clean validation bottles
- Seq:02
- Cat. No.:216-2288
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning