Orders LinkContactUsLinkComponent
VWR®, Fuel testing standards - gasoline
VWR®, Fuel testing standards - gasoline
Motor Octane Number standard, nominal value 86,9, ASTM D2700, 1 L
Katalog # 99129.290
Leverandør:   VWR Chemicals
VWR®, Fuel testing standards - gasoline
Katalog # 99129.290
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, Fuel testing standards - gasoline
VWR®, Fuel testing standards - gasoline
Motor Octane Number standard, nominal value 86,9, ASTM D2700, 1 L
CAS nummer:  
Price
6.500,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Fuel testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Octane Number.
  • Certified in accordance with ASTM / EN or IP test method protocols
  • Certified by Round Robin method using independent test laboratories
  • Ensures good inter-laboratory correlation
  • Supplied in tamper-evident, UN compliant security pack
Specifikationer
Specifikationer
  • Pakn:
    1 L
  • Beskrivelse:
    Motor Octane Number standard, nominal value 86,9
  • Testmetode:
    ASTM D2700
Specifikationer