VWR®, Fuel testing standards - gasoline
Motor Octane Number standard, nominal value 86,9, ASTM D2700, 1 L
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Fuel testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Octane Number.
- Certified in accordance with ASTM / EN or IP test method protocols
- Certified by Round Robin method using independent test laboratories
- Ensures good inter-laboratory correlation
- Supplied in tamper-evident, UN compliant security pack
Documents
Specifikationer
Specifikationer
- Pakn:1 L
- Beskrivelse:Motor Octane Number standard, nominal value 86,9
- Testmetode:ASTM D2700