Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Low Temperature viscosity standards

Standard with certified values at –24 °C, –24.5 °C, –25 °C, –25.5 °C and –26 °C, N400B, (–25 °C): 68176 mm²/s, Glass bottle, 500 mL
  99488.260
VWR®, Low Temperature viscosity standards
  99488.260
 :  
VWR®, Low Temperature viscosity standards
VWR®, Low Temperature viscosity standards

Standard with certified values at –24 °C, –24.5 °C, –25 °C, –25.5 °C and –26 °C, N400B, (–25 °C): 68176 mm²/s, Glass bottle, 500 mL

 :  
 
933,00 kr
   
1 * 500 mL
 

Low Temperature viscosity standards have certified data for kinematic viscosity, dynamic viscosity and density for various low temperatures.

  • Fully traceable to national standards
  • Ensures full compliance to ASTM and IP test method protocols
  • Tamper-evident security packaging

Low Temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.

 : Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.

 : The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.

 

 

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    Standard with certified values at –24 °C, –24.5 °C, –25 °C, –25.5 °C and –26 °C
  • Antal:
    N400B
  • Viskositet:
    (–25 °C): 68176 mm²/s