Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, Low Temperature viscosity standards

VWR®, Low Temperature viscosity standards

Standard with certified values at –25 °C, –30 °C, –35 °C and –40 °C, N14B, (–40 °C): 19605 mm²/s, Glass bottle, 500 mL
Katalog # 99485.260
Leverandør:   VWR Chemicals
VWR®, Low Temperature viscosity standards
Katalog # 99485.260
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, Low Temperature viscosity standards
VWR®, Low Temperature viscosity standards

Standard with certified values at –25 °C, –30 °C, –35 °C and –40 °C, N14B, (–40 °C): 19605 mm²/s, Glass bottle, 500 mL

CAS nummer:  
Price
933,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Low Temperature viscosity standards have certified data for kinematic viscosity, dynamic viscosity and density for various low temperatures.

  • Fully traceable to national standards
  • Ensures full compliance to ASTM and IP test method protocols
  • Tamper-evident security packaging

Low Temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.

Certifications: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.

Advarsel: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    Standard with certified values at –25 °C, –30 °C, –35 °C and –40 °C
  • Antal:
    N14B
  • Viskositet:
    (–40 °C): 19605 mm²/s

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.