Polysorbat 80, GPR RECTAPUR®
Plastic jerrican, 25 L
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Synonymer: Sorbitanmono(cis-9-octadecenoat), ethoxyliert
- CAS nummer:9005-65-6
- MDL nummer:MFCD00082107
- Merck Index:13,07664
- Kogende Pt:>100 °C (1013 hPa)
- Smeltende Pt:–21 °C
- Tæthed:1,062 g/cm³ (20 °C)
- Flash Pt:149 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- EINECS:500-019-9
Specifikationer
Specifikationstestresultater
- Identification:Passes test
- Density (20/4):1.070 - 1.090
- Hydroxyl value:65 - 80
- Saponification value:45 - 55
- Acid value:Max 2 mg KOH/g
- Water content:Max 3%
Specifikationer
- Pakning:Plastic jerrican
- Pakn:25 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



