Orders LinkContactUsLinkComponent
D3000 series particulate filters, Secure Click™
Secure Click™ particulate filter P2 R, D3125
  111-2606
 :   3M
D3000 series particulate filters, Secure Click™
  111-2606
 :  3M
D3000 series particulate filters, Secure Click™
D3000 series particulate filters, Secure Click™
Secure Click™ particulate filter P2 R, D3125
2.030,00 kr
1 * 20 Items
3M™ Secure Click™ D3000 Series Particulate Filters feature a new dual flow design for easier breathing.
  • New dual flow filters allow for easier breathing
  • Protects against particulates with nuisance level organic vapour/acid gas relief
  • Protection against ozone up to 10× occupational exposure limit
  • Quick, simple connection; simply align connections and push until an audible click is heard
  • Use with 3M™ Secure Click™ Reusable Respirators 800 Series
  • Swept-back design allows an enhanced field of view and comfort
  • Use for protection in hazardous environments
Secure Click™ Particulate Filter P3 R D3138 with Nuisance Level Organic Vapour Relief helps provide respiratory protection against exposure to certain particles. It also offers nuisance level odour relief from organic vapours that are below the OEL. Nuisance level refers to concentrations below OEL.
Secure Click™ Particulate Filter P3 R D3135 is CE approved for environments containing certain particles. It is a lightweight filter that combines protection with comfort.
Secure Click™ Particulate Filter P2 R D3128 is CE approved for environments containing certain particles. It can also be used for nuisance level organic vapour relief and for ozone protection up to 10× OEL. It is a lightweight filter that combines protection with comfort.
Secure Click™ Particulate Filter P2 R D3125 is CE approved for environments containing certain particles. It is a lightweight filter that combines protection with comfort.
These filters are commonly used for welding, brazing, torch cutting, metal pouring, and soldering. Inventory needs and training requirements of safety equipment are reduced because these filter works for many different applications. They are most commonly used in agriculture, construction, general manufacturing, pharmaceutical, and primary metal industries. These filters work with the 3M Secure Click™ Respirators 800 Series. The connection system works just like a seat belt. Simply align the connections and push until an audible click is heard. Used in pairs, the filters offer low breathing resistance and good balance when connected to the mask. They are designed to optimise the field of vision.
  • Beskrivelse:
    Secure Click™ particulate filter P2 R, D3125
  • Seq:
    04