Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Chitosan, powder

Chitosan, powder

Poly bottle, 100 g
Katalog # SPCMC1569-100GM
Leverandør:   Spectrum Chemical
Chitosan, powder
Katalog # SPCMC1569-100GM
Leverandør:  Spectrum Chemical
CAS nummer:  9012-76-4
Chitosan, powder

Poly bottle, 100 g

CAS nummer:  9012-76-4
Price
1.330,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Chitosan, Powder is a polysaccharide made by treating shrimp and other crustacean shells with the alkali sodium hydroxide. It can be used in agriculture as a seed treatment and to fight off fungal infections and is often used in the wine making industry as a chemical to prevent spoilage. In medicine it can be used in bandages to reduce bleeding and as an antibacterial agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chitosan can be used in agriculture as a seed treatment and biopesticide to fight off fungal infections and is often used in the wine making industry as a chemical to prevent spoilage. In medicine it can be used in bandages to reduce bleeding and as an antibacterial agent.

Synonymer:  Deacetylated chitin, Poly(D-glucosamine)
  • CAS nummer:9012-76-4
  • MDL nummer:MFCD00161512
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationstestresultater

  • Synonyms:
    β-(1,4)-2-Amino-2-deoxy-D-glucose; Deacetylated Chitin; Poly(D-glucosamine)
  • Form:
    Powder
  • Deacetylation:
    Min. 90.0 %
  • Moisture:
    Max. 10.0 %
  • Ash:
    Max. 5.0 %

Specifikationer

  • Pakning:
    Poly bottle
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.