Objectives for Rebel Microscopes
Plan achromat phase, N.A. 0,25, 10×, 10,8 mm
Katalog # ECHLRMAG0113
Leverandør: DISCOVER ECHO
Gå til:
Product Details & Documents
Specifikationer
- Forstørrelse:10×
- Beskrivelse:Plan achromat phase, N.A. 0,25
- Seq:25
- Arbejdsafstand:10,8 mm