Racks for Axygen® MultiRack tips
MultiRack empty rack, 5/pack, 1 pack/case, 10 µl extended length, 200 µl, 200 µl NX, 300 µl tips
Leverandør: Corning
Gå til:
Product Details & Documents
Documents
Specifikationer
- Beskrivelse:MultiRack empty rack, 5/pack, 1 pack/case
- Til brug med:10 µl extended length, 200 µl, 200 µl NX, 300 µl tips