Clips for conical ground joints
grøn, 24/29
Katalog # HECH42386024
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Clips made of POM for conical ground joints.
- Material does not scratch glass surfaces
- Resistant to temperatures up to +150 °C
Specifikationer
- Til NS:24/29
- Farve:grøn
- Seq:05