Clips for conical ground joints
blå, 19/26
Katalog # HECH42386019
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Clips made of POM for conical ground joints.
- Material does not scratch glass surfaces
- Resistant to temperatures up to +150 °C
Specifikationer
- Til NS:19/26
- Farve:blå
- Seq:04