Orders LinkContactUsLinkComponent
Safety faceshields, Shape™
Safety faceshields, Shape™
Shape™ safety faceshield, clear visor
Katalog # 111-2594
Leverandør:   JSP
Safety faceshields, Shape™
Katalog # 111-2594
Leverandør:  JSP
Safety faceshields, Shape™
Safety faceshields, Shape™
Shape™ safety faceshield, clear visor
Price
166,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
A compact, comfortable, and lightweight faceshield with superior optical quality.
  • Perfect optical quality
  • 2,2 mm polycarbonate visor
  • Wide field of vision
  • Adjustable wheel ratchet headband
  • Compatible with prescription glasses
The Shape™ Safety Faceshield features a moulded polycarbonate visor that provides superior optical quality with an exceptional field of vision.
Certifications: EN 166; EN 170.
Documents
Specifikationer
  • Beskrivelse:
    Shape™ safety faceshield, clear visor
  • Seq:
    02