Orders LinkContactUsLinkComponent
Nylon Wire and Cable Ties
Nylon Wire and Cable Ties
Nylon cable ties, width 3,5 mm, ufarvet, 14 cm
Katalog # BRDY196310
Leverandør:   Brady
Nylon Wire and Cable Ties
Katalog # BRDY196310
Leverandør:  Brady
Nylon Wire and Cable Ties
Nylon Wire and Cable Ties
Nylon cable ties, width 3,5 mm, ufarvet, 14 cm
Price
40,40 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Specifikationer
  • Farve:
    ufarvet
  • Beskrivelse:
    Nylon cable ties, width 3,5 mm
  • Længde:
    14 cm
  • Seq:
    02