VWR®, Wire baskets with two handles
540×255×60 mm
Catalog # 215-4230
Supplier: VWR Collection
Jump to:
Product Details & Documents
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
- Very good chemical and temperature resistance
- Two flip handles
- Base frame is stackable
Suitable for sterilisation of laboratory utensils.
Specifications
- B×D×H:540×255×60 mm