Orders LinkContactUsLinkComponent
VWR®, Wire baskets with two handles
VWR®, Wire baskets with two handles
540×255×60 mm
Catalog # 215-4230
Supplier:   VWR Collection
VWR®, Wire baskets with two handles
Catalog # 215-4230
Supplier:  VWR Collection
VWR®, Wire baskets with two handles
VWR®, Wire baskets with two handles
540×255×60 mm
Price
kr2,250.00
Log in for availability
1 * 1 Items
Product Details & Documents
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Two flip handles
  • Base frame is stackable
Suitable for sterilisation of laboratory utensils.
Specifications
  • B×D×H:
    540×255×60 mm