VWR®, Wire baskets with two handles
480×250×60 mm
215-4229
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
- Very good chemical and temperature resistance
- Two flip handles
- Base frame is stackable
Suitable for sterilisation of laboratory utensils.
- B×D×H:480×250×60 mm