Orders LinkContactUsLinkComponent
VWR®, Wire baskets with two handles
480×250×60 mm
  215-4229
VWR®, Wire baskets with two handles
  215-4229
VWR®, Wire baskets with two handles
VWR®, Wire baskets with two handles
480×250×60 mm
1.560,00 kr
1 * 1 Items
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Two flip handles
  • Base frame is stackable
Suitable for sterilisation of laboratory utensils.
  • B×D×H:
    480×250×60 mm