VWR®, Wire baskets with two handles
240×250×50 mm
Katalog # 215-4228
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
- Very good chemical and temperature resistance
- Two flip handles
- Base frame is stackable
Suitable for sterilisation of laboratory utensils.
Specifikationer
- B×D×H:240×250×50 mm