Orders LinkContactUsLinkComponent
VWR®, Wire baskets with two handles
VWR®, Wire baskets with two handles
240×250×50 mm
Katalog # 215-4228
Leverandør:   VWR Collection
VWR®, Wire baskets with two handles
Katalog # 215-4228
Leverandør:  VWR Collection
VWR®, Wire baskets with two handles
VWR®, Wire baskets with two handles
240×250×50 mm
Price
1.180,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Two flip handles
  • Base frame is stackable
Suitable for sterilisation of laboratory utensils.
Specifikationer
  • B×D×H:
    240×250×50 mm