Orders LinkContactUsLinkComponent
VWR®, Wire baskets
VWR®, Wire baskets
200×400×200 mm
Katalog # 215-4207
Leverandør:   VWR Collection
VWR®, Wire baskets
Katalog # 215-4207
Leverandør:  VWR Collection
VWR®, Wire baskets
VWR®, Wire baskets
200×400×200 mm
Price
2.180,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Spot-welded mesh 8×8×1 mm
  • Reinforced edges
Can be used for transport, storage or sterilisation of laboratory utensils.
Specifikationer
  • B×D×H:
    200×400×200 mm