VWR®, Wire baskets
140×140×140 mm
Katalog # 215-4202
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
- Very good chemical and temperature resistance
- Spot-welded mesh 8×8×1 mm
- Reinforced edges
Can be used for transport, storage or sterilisation of laboratory utensils.
Documents
Specifikationer
- B×D×H:140×140×140 mm