VWR®, Wire baskets
120×120×120 mm
Catalog # 215-4201
Supplier: VWR Collection
Jump to:
Product Details & Documents
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
- Very good chemical and temperature resistance
- Spot-welded mesh 8×8×1 mm
- Reinforced edges
Can be used for transport, storage or sterilisation of laboratory utensils.
Documents
Specifications
- B×D×H:120×120×120 mm