Orders LinkContactUsLinkComponent
Septa for open top caps, WHEATON®
Septa for open top caps, WHEATON®
Plain septa, autoclavable, PTFE faced semi-translucent silicone, 24 mm
Katalog # WHEAW240598A
Leverandør:   DWK Life Sciences
Septa for open top caps, WHEATON®
Katalog # WHEAW240598A
Leverandør:  DWK Life Sciences
Septa for open top caps, WHEATON®
Plain septa, autoclavable, PTFE faced semi-translucent silicone, 24 mm
Price
5.930,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Specifikationer
  • Ø:
    24 mm
  • Model:
    PTFE faced semi-translucent silicone
  • Beskrivelse:
    Plain septa, autoclavable
  • Seq:
    13