Septa for open top caps, WHEATON®
Plain septa, autoclavable, PTFE faced semi-translucent silicone, 24 mm
Katalog # WHEAW240598A
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Specifikationer
- Ø:24 mm
- Model:PTFE faced semi-translucent silicone
- Beskrivelse:Plain septa, autoclavable
- Seq:13