Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Eppendorf® Conical Tubes with SnapTec® Cap

Eppendorf quality (non sterile), ufarvet, 200 tubes (5 bags×40 tubes)
  525-1603
   Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
 :   EPPENDORF
Eppendorf® Conical Tubes with SnapTec® Cap
  525-1603
 :  EPPENDORF
   Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
Eppendorf® Conical Tubes with SnapTec® Cap
Eppendorf® Conical Tubes with SnapTec® Cap

Eppendorf quality (non sterile), ufarvet, 200 tubes (5 bags×40 tubes)

 
500,00 kr
   
1 * 200 Items
 

Eppendorf® Conical Tubes with SnapTec® caps offer secure sample containment for diverse applications. The innovative SnapTec® Cap ensures easy, secure sealing, making them ideal for molecular biology, genomics, and more. The conical design aids optimal sample recovery, enhancing efficiency and reliability in the lab.

  • SnapTec® caps allow effortless, secure sealing for sample integrity in molecular biology and genomics
  • Conical design enhances recovery, crucial for applications like PCR, DNA/RNA isolation, and centrifugation
  • Well-suited for various lab tasks, ensuring adaptability across experiments
  • Durable construction crafted for longevity, supporting applications like cell culture, protein purification, and more
  • Streamlines processes, benefitting applications such as qPCR, cloning, and sample storage
  • Available in 25 ml and SnapTec® 50 to 45 ml formats

Eppendorf® Conical Tubes with SnapTec® Caps redefine sample containment, catering to diverse lab applications. The innovative SnapTec® Cap ensures secure sealing for molecular biology and genomics, preserving sample integrity during storage. Versatile across lab tasks, they adapt to cell culture and protein purification. Crafted for durability, these tubes support longevity in various applications. Elevate scientific processes with Eppendorf®, offering trusted quality for versatile lab applications.

Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Material: PP
Maximum RCF: 17000×g

 : Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Sterile Grade can be downloaded from the Eppendorf website.

 
  • Pakket:
    200 tubes (5 bags×40 tubes)
  • Farve:
    ufarvet
  • Beskrivelse:
    Eppendorf quality (non sterile)
  • Seq:
    01
  • Cat. No.:
    525-1603
  • Operating temperature range:
    −86...+100 °C
  • Miljøcertificering:
    Renewable Energy Certificate, ISO 14001, ACT Label
  • Mål:
    Ø 30 mm
  • Environmentally Preferable:
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Autoklaverbare:
    When open, 121 °C, 20 min
Accessories

 

  •  
    Beskrivelse
     
     
  • EPPE0030119.860
    Single tube stand, for 25 and 50 ml tubes
       
    1 * 4 Items - 180,00 kr

 

  •  
    Beskrivelse
     
     
  • EPPE0030119.835
    12 positions, 6 for 5,0 and 15 ml tubes; 6 for 25 and 50 ml tubes
       
    1 * 2 Items - 390,00 kr

 

  •  
    Beskrivelse
     
     
  • EPPE5353040.156
    Tube holder 25/50 ml, for 4×25/50 ml conical tubes
       
    1 * 1 Items - 850,00 kr

 

  •  
    Beskrivelse
    Rum
    Til
     
     
  • EPPE0030140.729
    Hole Ø: 30,3 mm
    3×3
    Tubes 25 ml
       
    1 * 2 Items - 180,00 kr

 

  •  
    Beskrivelse
     
     
  • EPPE5820733.004
    Adapter, for 1 Eppendorf conical tube 25 ml with snap cap 25 ml snap cap, 6 peices.
       
    1 * 6 Items - 260,00 kr

 

  •  
    Beskrivelse
     
     
  • EPPE5365000.109
    Adapter 25 ml for SmartBlock 50 ml - set consisting of 4 adapters and removal tool
       
    1 * 1 Each - 1.310,00 kr

 

  •  
    Beskrivelse
     
     
  • EPPE5820734.000
    Adapter, for 1 Eppendorf conical tube 25 ml with screw cap 25 ml screw cap, 6 pc
       
    1 * 6 Items - 260,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.