Body lotion, Lindesa®
Body lotion, Lindesa®, 1000 ml tube
Katalog # FAWE13640-002
Leverandør: FAWECO HAUTSCHUTZ
Gå til:
Product Details & Documents
Barrier cream and care cream with beeswax is designed to care for skin exposed to minor contamination, and is also suitable for after-work care.
- Lightly greasy, absorbs quickly
- Special care ingredients improve the skin's elasticity and suppleness
- Does not affect the sense of touch
- Emulsion type: O/W, neutral pH level, silicone-free, scented
The cream strengthens the skin's immunity, prevents skin damage from exposure to external contamination, and has a soothing effect on irritated skin. It smooths and nourishes without any long-lasting, greasy film.
Caution: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifikationer
- Beskrivelse:Body lotion, Lindesa®, 1000 ml tube
- Seq:03