Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
Product Details & Documents
PIPETMAN®, recognized as the pipetting standard for its reliability and proven durability, is a fully adjustable, air-displacement pipette that uses disposable tips. PIPETMAN combines legendary accuracy, precision, and durability with easy pipetting and handling. The pipettes come in a large range of volumes with reduced pipetting forces for hours of comfortable pipetting.
- Legendary performance
- Easy pipetting
- Built-to-last and convenient
- Simple to maintain
Accurate and precise, PIPETMAN air-displacement pipettes are recognized as one of the world standard. They come in a large range of volumes, including single and multichannel models, allowing you to handle various sample types with confidence. PIPETMAN feels like PIPETMAN® Classic but with lower forces for enhanced pipetting habits with a standard pipette. Made of PVDF and stainless steel, PIPETMAN offers premium durability. Built-to-last, it is the economical solution for years of pipetting. Routine cleaning and maintenance are all that is required to keep PIPETMAN in top condition for years. The PIPETMAN design allows for easy access to user-serviceable parts, along with a fully autoclavable tip holder and ejector.
Bestillingsoplysninger: Models equipped with a tip ejector are only sold with a stainless steel tip ejector. Plastic tip ejectors are available in the accessories.
Specifikationer
- Farvekode:grøn
- Volumen:30 - 300 µl
- Beskrivelse:P8x300
- Seq:04
- Nøjagtighed:±1,00 - ±3,00 µl
- Usikkerhed:≤ 0,35 - ≤ 1,00 µl
- Til pipettespidser:D200 / DF200 / D300 / DF300
Accessories
Produktinformation
- Katalog #FarveTilgængelighedPrice
- Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)612-1816hvidLog ind for tilgængelighed1 * 1 Items - 939,96 kr
Produktinformation
- Katalog #FarveTilgængelighedPrice
- Angled pipette stand, TRIO™, for 3 Gilson pipettes (except Pipetman Concept®)Angled pipette stand, TRIO™, for 3 Gilson pipettes (except Pipetman Concept®)GILSF161405gennemsigtigLog ind for tilgængelighed1 * 1 Items - 828,06 kr
Produktinformation
- Katalog #FarveTilgængelighedPrice
- Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screwsPipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screwsGILSF161406hvidLog ind for tilgængelighed1 * 1 Items - 141,74 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Plastic ejector for P20
Produktinformation
- Katalog #TilgængelighedPrice
- Plastic ejector for P2/P10 with adapterPlastic ejector for P2/P10 with adapterGILSF107027Log ind for tilgængelighed1 * 1 Items - 117,87 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Plastic ejector for P1000
Produktinformation
- Katalog #TilgængelighedPrice
- Plastic ejector for P200
Produktinformation
- Katalog #TilgængelighedPrice
- Plastic ejector for P100