ESC (Easy Surface Checking) swabs
ESC swab Maximum Recovery Diluent, 100 tubes×5 ml
Katalog # 85610.
Leverandør: LIOFILCHEM
Gå til:
Product Details & Documents
The ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
- Pre-filled swab device
- Rayon bud
- Wide range of diluents
- Leak proof closure
- Transport and storage at room temperature
Certifications: Swabs conform to international standards (ISO 18593, ISO 17604).
Specifikationer
- Beskrivelse:ESC swab Maximum Recovery Diluent, 100 tubes×5 ml
- Seq:11