Orders LinkContactUsLinkComponent
Labels for BBP33 printers, type B-424
Labels for BBP33 printers, type B-424
Printer labels for general identification, B-424, Papir, Permanent, gummibaseret, Thermal transfer, hvid, BBP33, 69,9×31,8 mm
  BRDY133964
 :   Brady
Labels for BBP33 printers, type B-424
  BRDY133964
 :  Brady
Labels for BBP33 printers, type B-424
Labels for BBP33 printers, type B-424
Printer labels for general identification, B-424, Papir, Permanent, gummibaseret, Thermal transfer, hvid, BBP33, 69,9×31,8 mm
292,00 kr
1 * 1.000 Items
These paper labels are used for general identification and have a matt finish.
  • Printer:
    BBP33
  • Printing method:
    Thermal transfer
  • Model:
    B-424
  • Farve:
    hvid
  • Beskrivelse:
    Printer labels for general identification
  • Dimensioner:
    69,9×31,8 mm
  • Materiale:
    Papir
  • Seq:
    04
  • Klæbestof:
    Permanent, gummibaseret