Labels for BBP33 printers, type B-486
Metalised printer labels, B-486, Polyester, Permanent, gummibaseret, Thermal transfer, sølvfarvet, BBP33, 50,8×12,7 mm
Katalog # BRDY133954
Leverandør: Brady
Gå til:
Product Details & Documents
Metallised polyester labels for product labelling, asset tracking and general industrial ID.
- Matte, light gray material has a permanent rubber-based adhesive and is thermal transfer printable
- Ideal for serial number plates and rating plates with name-plate quality and metal-looking finish
- Service temperature from –40 to +120 °C
Certifications: UL approved, CSA accepted.
Specifikationer
- Printer:BBP33
- Printing method:Thermal transfer
- Model:B-486
- Farve:sølvfarvet
- Beskrivelse:Metalised printer labels
- Dimensioner:50,8×12,7 mm
- Materiale:Polyester
- Seq:03
- Klæbestof:Permanent, gummibaseret