Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ethidiumbromid ≥98% for biotechnology

Ethidiumbromid ≥98% for biotechnology

100 g
Katalog # 0492-100G
Sundhed
Giftig
Leverandør:   VWR Chemicals
Ethidiumbromid ≥98% for biotechnology
Katalog # 0492-100G
Leverandør:  VWR Chemicals
CAS nummer:  1239-45-8
Ethidiumbromid ≥98% for biotechnology
Ethidiumbromid ≥98% for biotechnology

100 g

CAS nummer:  1239-45-8
Price
14.780,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Fluorescent stain for visualizing nucleic acids.

At a concentration of 0.5 μg/ml, ethidium bromide is useful in agarose or acrylamide gels for band detection. It is used to detect as little as 10 ng of nucleic acid. At higher concentrations (0.5 to 1.0 μg/ml), it may be used to facilitate purification of DNA in a cesium chloride gradient.

For maximum convenience, VWR offers ethidium bromide in various forms, including ready-to-use solutions and powdered forms. Each is packaged in shatter-resistant packaging for added safety. VWR’s ethidium bromide dropper bottle offers the ultimate convenience and safety. Simply dispense one drop of ethidium bromide into 50ml of agarose gel or running buffer (TBE or TAE) or staining solution.

Synonymer:  3,8-Diamino-5-ethyl-6-phenylphenantridiniumbromid
  • CAS nummer:1239-45-8
  • Formel:C₂₁H₂₀BrN₃
  • MDL nummer:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Kogende Pt:72 °C (0,1 Torr)
  • Smeltende Pt:261…264 °C
  • Tæthed:1,01 g/cm³ (20 °C)
  • Flash Pt:<100 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:214-984-6
  • FN:2811
  • ADR:6.1,I

Documents

Specifikationer

Specifikationstestresultater

  • DNase:
    None Detected
  • Loss on Drying:
    ≤5%
  • Protease:
    None Detected
  • Purity (Dry Basis):
    ≥98%
  • RNase:
    None

Specifikationer

  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.