Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat CC, N.A. 0,95, 40×, 0,13 - 0,16 mm
Catalog # ECHLRMAG9404
Supplier:   DISCOVER ECHO
Objectives for Rebel Microscopes
Catalog # ECHLRMAG9404
Supplier:  DISCOVER ECHO
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Plan apochromat CC, N.A. 0,95, 40×, 0,13 - 0,16 mm
Price
25.850,00 kr
Log in for availability
1 * 1 Items
Specifications
  • Forstørrelse:
    40×
  • Beskrivelse:
    Plan apochromat CC, N.A. 0,95
  • Seq:
    01
  • Arbejdsafstand:
    0,13 - 0,16 mm