Orders LinkContactUsLinkComponent
Handheld digital microscopes, Dino-Lite Wireless
Handheld digital microscopes, Dino-Lite Wireless
Wireless microscope, WF4115ZT, 20 - 220×, 1,3 MP, -
Katalog # IDCPWF4115ZT
Leverandør:   Dino-Lite / IDCP B.V.
Handheld digital microscopes, Dino-Lite Wireless
Katalog # IDCPWF4115ZT
Leverandør:  Dino-Lite / IDCP B.V.
Handheld digital microscopes, Dino-Lite Wireless
Handheld digital microscopes, Dino-Lite Wireless
Wireless microscope, WF4115ZT, 20 - 220×, 1,3 MP, -
Price
7.140,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These microscopes from the Dino-Lite Universal series work wired via USB or wireless with the integrated WF-20 Wireless adapter.
  • Wireless and wired in one device
  • Work with different operating systems: iOS, Android or Windows
  • USB 2.0, enhanced 1,3 MP EDGE sensor
  • Integrated polariser greatly reduces glare on reflective surfaces
  • Extensive measurement functionality
  • Flexible LED control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Ideal for inspections in hard-to-reach places in factories, in the field, in workshops or in greenhouses.
Specifikationer
  • Forstørrelse:
    20 - 220×
  • Model:
    WF4115ZT
  • Opløsning:
    1,3 MP
  • Beskrivelse:
    Wireless microscope
  • Seq:
    01
  • Yderligere funktioner:
    -
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm
    Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm
    630-2517
    Log ind for tilgængelighed
    1 * 1 Items - 2.540,00 kr
Produktinformation
  • Katalog #
    B×D×H
    Tilgængelighed
    Price
  • Tabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX
    Tabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX
    IDCPRK-06A
    150×220×270 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.880,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Foot pedal, enables users to take a picture with their foot
    Foot pedal, enables users to take a picture with their foot
    630-1821
    Log ind for tilgængelighed
    1 * 1 Items - 472,00 kr