Handheld digital microscopes, Dino-Lite Wireless
Wireless microscope, WF4915ZT, 20 - 220×, 1,3 MP, AMR / EDOF / EDR
IDCPWF4915ZT
These microscopes from the Dino-Lite Universal series work wired via USB or wireless with the integrated WF-20 Wireless adapter.
- Wireless and wired in one device
- Work with different operating systems: iOS, Android or Windows
- USB 2.0, enhanced 1,3 MP EDGE sensor
- Integrated polariser greatly reduces glare on reflective surfaces
- Extensive measurement functionality
- Flexible LED control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Ideal for inspections in hard-to-reach places in factories, in the field, in workshops or in greenhouses.
- Forstørrelse:20 - 220×
- Model:WF4915ZT
- Opløsning:1,3 MP
- Beskrivelse:Wireless microscope
- Seq:05
- Yderligere funktioner:AMR / EDOF / EDR
Accessories
- Beskrivelse
- 630-2517Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm1 * 1 Items - 2.540,00 kr
- BeskrivelseB×D×H
- IDCPRK-06ATabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX150×220×270 mm1 * 1 Items - 1.880,00 kr
- Beskrivelse