Orders LinkContactUsLinkComponent
LED excitation modules
LED excitation modules
LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
Katalog # MOTI1101011400231
Leverandør:   MOTIC
LED excitation modules
Katalog # MOTI1101011400231
Leverandør:  MOTIC
LED excitation modules
LED excitation modules
LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube
Price
11.930,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    LED excitation module TRITC, consisting of 525 nm LED lamp and TRITC filter cube