Orders LinkContactUsLinkComponent
LED excitation modules
LED excitation modules
LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
Katalog # MOTI1101011400251
Leverandør:   MOTIC
LED excitation modules
Katalog # MOTI1101011400251
Leverandør:  MOTIC
LED excitation modules
LED excitation modules
LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
Price
13.700,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube