LED excitation modules
LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
Katalog # MOTI1101011400251
Leverandør: MOTIC
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube