Orders LinkContactUsLinkComponent
Conductivity standards, Supelco®
Conductivity standards, Supelco®
Conductivity standard, 75 µmhos/cm, 500 mL
Catalog # COND75-500ML
Supplier:   Merck
Conductivity standards, Supelco®
Catalog # COND75-500ML
Supplier:  Merck
CAS Number:  
Conductivity standards, Supelco®
Conductivity standard, 75 µmhos/cm, 500 mL
CAS Number:  
Price
628,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents
100 µmhos/cm = 100 µS/cm
Certifications: These certified reference materials (CRMs) ares produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Documents
Specifications
Specifications
  • Pakn:
    500 mL
  • Beskrivelse:
    Conductivity standard
  • Konduktivitet:
    75 µmhos/cm