Orders LinkContactUsLinkComponent
Conductivity standards, Supelco®
Conductivity standard, 147 µmhos/cm, 500 mL
Katalog # COND147-500ML
Leverandør:   Merck
Conductivity standards, Supelco®
Katalog # COND147-500ML
Leverandør:  Merck
CAS nummer:  
Conductivity standards, Supelco®
Conductivity standard, 147 µmhos/cm, 500 mL
CAS nummer:  
Price
735,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
100 µmhos/cm = 100 µS/cm
 : These certified reference materials (CRMs) ares produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Specifikationer
Specifikationer
  • Pakn:
    500 mL
  • Beskrivelse:
    Conductivity standard
  • Konduktivitet:
    147 µmhos/cm