Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Nicotine solution, NMR reference standard, Supelco®

Nicotine solution, NMR reference standard, Supelco®

Nicotine solution, 20 mg/mL in acetone-d6 (99.9 atom % D), TMS 0.05 %, CFCl3 0.4 %, NMR reference standard, NMR tube size 5 mm x 8 in., 10
Katalog # 613223-10X8IN
Leverandør:   Merck
Nicotine solution, NMR reference standard, Supelco®
Katalog # 613223-10X8IN
Leverandør:  Merck
CAS nummer:  
Nicotine solution, NMR reference standard, Supelco®

Nicotine solution, 20 mg/mL in acetone-d6 (99.9 atom % D), TMS 0.05 %, CFCl3 0.4 %, NMR reference standard, NMR tube size 5 mm x 8 in., 10

CAS nummer:  
Price
31.390,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Specifikationer

Specifikationer

  • Koncentration:
    20 mg/mL in acetone-d6 (99.9 atom % D), TMS 0.05 %, CFCl3 0.4 %
  • Pakn:
    10
  • Standard:
    NMR reference standard
  • Beskrivelse:
    Nicotine solution
  • Pk Info:
    NMR tube size 5 mm x 8 in.
  • Cat. No.:
    613223-10X8IN
  • Beskrivelse:
    Nicotine solution

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.