Nicotine solution, NMR reference standard, Supelco®
Nicotine solution, 2 mg/mL in acetonitrile-d3 (99.8 atom % D), TMS 0.05 %, NMR reference standard, NMR tube size 5 mm x 8 in., 10
Katalog # 612049-10X8IN
Leverandør: Merck
Nicotine solution, NMR reference standard, Supelco®
Katalog # 612049-10X8IN
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Koncentration:2 mg/mL in acetonitrile-d3 (99.8 atom % D), TMS 0.05 %
- Pakn:10
- Standard:NMR reference standard
- Beskrivelse:Nicotine solution
- Pk Info:NMR tube size 5 mm x 8 in.
- Cat. No.:612049-10X8IN
- Beskrivelse:Nicotine solution
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



