Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Nicotine solution, NMR reference standard, Supelco®

Nicotine solution, 2 mg/mL in chloroform-d (99.8 atom % D), TMS 0.05 %, NMR reference standard, NMR tube size 5 mm x 8 in., 10
Katalog # 612022-10X8IN
Leverandør:   Merck
Nicotine solution, NMR reference standard, Supelco®
Katalog # 612022-10X8IN
Leverandør:  Merck
CAS nummer:  
Nicotine solution, NMR reference standard, Supelco®

Nicotine solution, 2 mg/mL in chloroform-d (99.8 atom % D), TMS 0.05 %, NMR reference standard, NMR tube size 5 mm x 8 in., 10

CAS nummer:  
Price
20.990,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Specifikationer

Specifikationer

  • Koncentration:
    2 mg/mL in chloroform-d (99.8 atom % D), TMS 0.05 %
  • Pakn:
    10
  • Standard:
    NMR reference standard
  • Beskrivelse:
    Nicotine solution
  • Pk Info:
    NMR tube size 5 mm x 8 in.
  • Cat. No.:
    612022-10X8IN
  • Beskrivelse:
    Nicotine solution

Specifikationer