Nicotine solution, NMR reference standard, Supelco®
Nicotine solution, 2 mg/mL in benzene-d6 (99.6 atom % D), TMS 0.05 %, NMR reference standard, NMR tube size 5 mm x 8 in., 10
Catalog # 612014-10X8IN
Supplier: Merck
Nicotine solution, NMR reference standard, Supelco®
Catalog # 612014-10X8IN
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Koncentration:2 mg/mL in benzene-d6 (99.6 atom % D), TMS 0.05 %
- Pakn:10
- Standard:NMR reference standard
- Beskrivelse:Nicotine solution
- Pk Info:NMR tube size 5 mm x 8 in.
- Cat. No.:612014-10X8IN
- Beskrivelse:Nicotine solution
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



