Supelco® Organic Reference Standard, Acetaldehyde-2,4-DNPH solution
Acetaldehyde-2,4-DNPH solution, Ampule of 5×1 ml, Certified reference material, Ampule of 5×1 ml, 5
Leverandør: Merck
Supelco® Organic Reference Standard, Acetaldehyde-2,4-DNPH solution
Katalog # CRM4M7340
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
- GHS signal: GHS02, GHS07
Advarsel: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Koncentration:Ampule of 5×1 ml
- Pakn:5
- Standard:Certified reference material
- Beskrivelse:Acetaldehyde-2,4-DNPH solution
- Pk Info:Ampule of 5×1 ml
- Cat. No.:CRM4M7340
- M-Clarity quality level:MQ100
- CAS-nr:1019-57-4
- Molekylvægt:224,17
- Fareklassebeskrivelse:H225 - H302 + H312 + H332 - H319
- Flammepunkt:2,0 °C - Closed cup
- Opbevaring betingelser:2…8 °C
- Synonym(s):Acetaldehyde-2,4-dinitrophenylhydrazone solution
- Shipping conditions:WICE
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



