Supelco® Organic Reference Standard, Acetaldehyde-2,4-DNPH solution
Acetaldehyde-2,4-DNPH solution, 1000 µg/ml in acetonitrile, Certified reference material, Ampule of 1 ml, 1 mL
Katalog # CRM47340
Leverandør: Merck
Supelco® Organic Reference Standard, Acetaldehyde-2,4-DNPH solution
Katalog # CRM47340
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
- GHS signal: GHS02, GHS07
Advarsel: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Koncentration:1000 µg/ml in acetonitrile
- Pakn:1 mL
- Standard:Certified reference material
- Beskrivelse:Acetaldehyde-2,4-DNPH solution
- Pk Info:Ampule of 1 ml
- Cat. No.:CRM47340
- M-Clarity quality level:MQ100
- CAS-nr:1019-57-4
- Molekylvægt:224,17
- Fareklassebeskrivelse:H225 - H302 + H312 + H332 - H319
- Flammepunkt:2,0 °C - Closed cup
- Opbevaring betingelser:2…8 °C
- Synonym(s):Acetaldehyde-2,4-dinitrophenylhydrazone solution
- Shipping conditions:WICE
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



