Orders LinkContactUsLinkComponent
Water baths, PURA™
Water baths, PURA™
PURA 14, 18...99 °C, ±0,15 °C, 1,8 kW, 2 - 14 L, 330×270×140 mm, 420×350×220 mm
Katalog # 462-0627
Leverandør:   JULABO GmbH
Water baths, PURA™
Katalog # 462-0627
Leverandør:  JULABO GmbH
Water baths, PURA™
Water baths, PURA™
PURA 14, 18...99 °C, ±0,15 °C, 1,8 kW, 2 - 14 L, 330×270×140 mm, 420×350×220 mm
Price
8.510,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PURA stainless steel water baths are functional and reliable for everyday operations due to their high material and component quality. The easy to use PURA water baths offer a number of functions and features that enable efficient use in the laboratory.
  • Intuitive, straightforward menu and bright, easy to read LED display
  • Splash-proof protected power switch
  • Integral drain screw (except PURA 4) and integrated timer
  • Built-in dry running protection
  • No interfering elements in the bath
  • Optical and acoustic alarm
  • Non slip rubber feet and ergonomic recessed grips
Units feature specially designed edge surfaces, which automatically drain condensed fluid back into the bath. Supplied with a removable platform which allows sample containers placed on the platform to be fully immersed and thus optimally tempered. PURA water baths are designed to enable natural circulation within the tank, all without a pump, guarantying homogeneous temperatures.
Delivery information: Supplied with platform but without lids, a wide range of optional accessories are available. Note PURA™ 4, 10, 14, 22 and 30 baths accommodate a maximum of 1, 2, 3, 5 and 7 optional test tube racks respectively.
Documents
Specifikationer
  • Opvarmnings kapacitet:
    1,8 kW
  • Temperaturområde:
    18...99 °C
  • Kapacitet:
    2 - 14 L
  • Seq:
    003
  • B×D×H:
    420×350×220 mm
  • Bad B×D×H:
    330×270×140 mm
  • Temp. stabilitet:
    ±0,15 °C
  • Model:
    PURA 14
Accessories
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Flat bath lid, ring sets with 3 openings, 190 mm Ø
    Flat bath lid, ring sets with 3 openings, 190 mm Ø
    JULA9970577
    PURA™ 30
    Log ind for tilgængelighed
    1 * 1 Items - 2.350,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Hule PP-kugler, 20 mm Ø
    Hule PP-kugler, 20 mm Ø
    461-3619
    Åbne varmebade, termostatbade, vandbade og rystevandbade
    Log ind for tilgængelighed
    1 * 1.000 Items - 1.090,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Liquid level/cooling set
    Liquid level/cooling set
    462-0499
    PURA™ 10, PURA™ 14, PURA™ 22, PURA™30, SW22, SW23
    Log ind for tilgængelighed
    1 * 1 Items - 2.300,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Flat bath lid, ring sets with 6 openings, 115 mm Ø
    Flat bath lid, ring sets with 6 openings, 115 mm Ø
    JULA9970574
    PURA™ 22
    Log ind for tilgængelighed
    1 * 1 Items - 2.840,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Flat bath lid, ring sets with 2 openings, 190 mm Ø
    Flat bath lid, ring sets with 2 openings, 190 mm Ø
    JULA9970575
    PURA™ 22
    Log ind for tilgængelighed
    1 * 1 Items - 1.930,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Flat bath lid, ring sets with 8 openings, 92 mm Ø
    Flat bath lid, ring sets with 8 openings, 92 mm Ø
    JULA9970573
    PURA™ 22
    Log ind for tilgængelighed
    1 * 1 Items - 3.390,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Transparent lift up lid, for use up to +100 °C
    Transparent lift up lid, for use up to +100 °C
    461-1449
    PURA™ 4
    Log ind for tilgængelighed
    1 * 1 Items - 1.270,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Transparent lift up lid, for use up to +100 °C
    Transparent lift up lid, for use up to +100 °C
    461-1450
    PURA™ 10
    Log ind for tilgængelighed
    1 * 1 Items - 1.590,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Cooling coil for working near ambient temperature
    Cooling coil for working near ambient temperature
    JULA9970534
    PURA™ 10, PURA™ 14
    Log ind for tilgængelighed
    1 * 1 Items - 1.700,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Cooling coil for working near ambient temperature
    Cooling coil for working near ambient temperature
    JULA9970536
    PURA™ 22, PURA™ 30
    Log ind for tilgængelighed
    1 * 1 Items - 2.010,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Flat bath lid, ring sets with 4 openings, 92 mm Ø
    Flat bath lid, ring sets with 4 openings, 92 mm Ø
    JULA9970570
    PURA™ 10
    Log ind for tilgængelighed
    1 * 1 Items - 1.850,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Transparent lift up lid, for use up to +100 °C
    Transparent lift up lid, for use up to +100 °C
    461-1453
    PURA™ 30
    Log ind for tilgængelighed
    1 * 1 Items - 2.500,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Plastic test tube rack for 21 tubes with 30 mm Ø
    Plastic test tube rack for 21 tubes with 30 mm Ø
    461-0943
    B13, B17, B19, B27, B33, BT19, BT27, PURA™
    Log ind for tilgængelighed
    1 * 1 Items - 481,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Plastic test tube rack for 90 tubes with 12/13 mm Ø, height 75 mm
    Plastic test tube rack for 90 tubes with 12/13 mm Ø, height 75 mm
    461-0942
    B13, B17, B19, B27, B33, BT19, BT27, PURA™
    Log ind for tilgængelighed
    1 * 1 Items - 481,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Flat bath lid, ring sets with 1 opening, 190 mm Ø
    Flat bath lid, ring sets with 1 opening, 190 mm Ø
    JULA9970571
    PURA™ 10
    Log ind for tilgængelighed
    1 * 1 Items - 1.140,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Plastic test tube rack for 60 tubes with 17 mm Ø, height 100 mm
    Plastic test tube rack for 60 tubes with 17 mm Ø, height 100 mm
    461-0941
    B13, B17, B19, B27, B33, BT19, BT27, PURA™
    Log ind for tilgængelighed
    1 * 1 Items - 481,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Flat bath lid, ring sets with 10 openings, 115 mm Ø
    Flat bath lid, ring sets with 10 openings, 115 mm Ø
    JULA9970576
    PURA™ 30
    Log ind for tilgængelighed
    1 * 1 Items - 4.700,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Transparent lift up lid, for use up to +100 °C
    Transparent lift up lid, for use up to +100 °C
    461-1451
    PURA™ 14
    Log ind for tilgængelighed
    1 * 1 Items - 1.700,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Flat bath lid, ring sets with 6 openings, 92 mm Ø
    Flat bath lid, ring sets with 6 openings, 92 mm Ø
    JULA9970572
    PURA™ 14
    Log ind for tilgængelighed
    1 * 1 Items - 2.630,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Transparent lift up lid, for use up to +100 °C
    Transparent lift up lid, for use up to +100 °C
    461-1452
    PURA™ 22
    Log ind for tilgængelighed
    1 * 1 Items - 2.070,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Drain tap with tubing, ID 8 mm
    Drain tap with tubing, ID 8 mm
    JULA8970453
    PURA™ 10, PURA™ 14, PURA™ 22, PURA™ 30, SE-12, SE-26, SE-6, SE-Z, SL-12, SL-26, SL-6, SW22, SW23, HE-4, HL-4
    Log ind for tilgængelighed
    1 * 1 Items - 882,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Stainless steel hygiene insert for dentistry work
    Stainless steel hygiene insert for dentistry work
    JULA9970532
    PURA™ 4
    Log ind for tilgængelighed
    1 * 1 Items - 850,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Stents lifter for dentistry work
    Stents lifter for dentistry work
    461-1026
    PURA™ 4
    Log ind for tilgængelighed
    1 * 1 Items - 913,00 kr