Water baths, PURA™
Product Details & Documents
PURA stainless steel water baths are functional and reliable for everyday operations due to their high material and component quality. The easy to use PURA water baths offer a number of functions and features that enable efficient use in the laboratory.
- Intuitive, straightforward menu and bright, easy to read LED display
- Splash-proof protected power switch
- Integral drain screw (except PURA 4) and integrated timer
- Built-in dry running protection
- No interfering elements in the bath
- Optical and acoustic alarm
- Non slip rubber feet and ergonomic recessed grips
Units feature specially designed edge surfaces, which automatically drain condensed fluid back into the bath. Supplied with a removable platform which allows sample containers placed on the platform to be fully immersed and thus optimally tempered. PURA water baths are designed to enable natural circulation within the tank, all without a pump, guarantying homogeneous temperatures.
Delivery information: Supplied with platform but without lids, a wide range of optional accessories are available. Note PURA™ 4, 10, 14, 22 and 30 baths accommodate a maximum of 1, 2, 3, 5 and 7 optional test tube racks respectively.
Documents
JULABO PURA Water Baths
Videos
Specifikationer
- Opvarmnings kapacitet:1,8 kW
- Temperaturområde:18...99 °C
- Kapacitet:2 - 14 L
- Seq:003
- B×D×H:420×350×220 mm
- Bad B×D×H:330×270×140 mm
- Temp. stabilitet:±0,15 °C
- Model:PURA 14
Accessories
Produktinformation
- Katalog #TilTilgængelighedPrice
- Flat bath lid, ring sets with 3 openings, 190 mm ØFlat bath lid, ring sets with 3 openings, 190 mm ØJULA9970577PURA™ 30Log ind for tilgængelighed1 * 1 Items - 2.350,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Hule PP-kugler, 20 mm ØHule PP-kugler, 20 mm Ø461-3619Åbne varmebade, termostatbade, vandbade og rystevandbadeLog ind for tilgængelighed1 * 1.000 Items - 1.090,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Liquid level/cooling setLiquid level/cooling set462-0499PURA™ 10, PURA™ 14, PURA™ 22, PURA™30, SW22, SW23Log ind for tilgængelighed1 * 1 Items - 2.300,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Flat bath lid, ring sets with 6 openings, 115 mm ØFlat bath lid, ring sets with 6 openings, 115 mm ØJULA9970574PURA™ 22Log ind for tilgængelighed1 * 1 Items - 2.840,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Flat bath lid, ring sets with 2 openings, 190 mm ØFlat bath lid, ring sets with 2 openings, 190 mm ØJULA9970575PURA™ 22Log ind for tilgængelighed1 * 1 Items - 1.930,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Flat bath lid, ring sets with 8 openings, 92 mm ØFlat bath lid, ring sets with 8 openings, 92 mm ØJULA9970573PURA™ 22Log ind for tilgængelighed1 * 1 Items - 3.390,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Transparent lift up lid, for use up to +100 °CTransparent lift up lid, for use up to +100 °C461-1449PURA™ 4Log ind for tilgængelighed1 * 1 Items - 1.270,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Transparent lift up lid, for use up to +100 °CTransparent lift up lid, for use up to +100 °C461-1450PURA™ 10Log ind for tilgængelighed1 * 1 Items - 1.590,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Cooling coil for working near ambient temperatureCooling coil for working near ambient temperatureJULA9970534PURA™ 10, PURA™ 14Log ind for tilgængelighed1 * 1 Items - 1.700,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Cooling coil for working near ambient temperatureCooling coil for working near ambient temperatureJULA9970536PURA™ 22, PURA™ 30Log ind for tilgængelighed1 * 1 Items - 2.010,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Flat bath lid, ring sets with 4 openings, 92 mm ØFlat bath lid, ring sets with 4 openings, 92 mm ØJULA9970570PURA™ 10Log ind for tilgængelighed1 * 1 Items - 1.850,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Transparent lift up lid, for use up to +100 °CTransparent lift up lid, for use up to +100 °C461-1453PURA™ 30Log ind for tilgængelighed1 * 1 Items - 2.500,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Plastic test tube rack for 21 tubes with 30 mm ØPlastic test tube rack for 21 tubes with 30 mm Ø461-0943B13, B17, B19, B27, B33, BT19, BT27, PURA™Log ind for tilgængelighed1 * 1 Items - 481,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Plastic test tube rack for 90 tubes with 12/13 mm Ø, height 75 mmPlastic test tube rack for 90 tubes with 12/13 mm Ø, height 75 mm461-0942B13, B17, B19, B27, B33, BT19, BT27, PURA™Log ind for tilgængelighed1 * 1 Items - 481,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Flat bath lid, ring sets with 1 opening, 190 mm ØFlat bath lid, ring sets with 1 opening, 190 mm ØJULA9970571PURA™ 10Log ind for tilgængelighed1 * 1 Items - 1.140,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Plastic test tube rack for 60 tubes with 17 mm Ø, height 100 mmPlastic test tube rack for 60 tubes with 17 mm Ø, height 100 mm461-0941B13, B17, B19, B27, B33, BT19, BT27, PURA™Log ind for tilgængelighed1 * 1 Items - 481,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Flat bath lid, ring sets with 10 openings, 115 mm ØFlat bath lid, ring sets with 10 openings, 115 mm ØJULA9970576PURA™ 30Log ind for tilgængelighed1 * 1 Items - 4.700,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Transparent lift up lid, for use up to +100 °CTransparent lift up lid, for use up to +100 °C461-1451PURA™ 14Log ind for tilgængelighed1 * 1 Items - 1.700,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Flat bath lid, ring sets with 6 openings, 92 mm ØFlat bath lid, ring sets with 6 openings, 92 mm ØJULA9970572PURA™ 14Log ind for tilgængelighed1 * 1 Items - 2.630,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Transparent lift up lid, for use up to +100 °CTransparent lift up lid, for use up to +100 °C461-1452PURA™ 22Log ind for tilgængelighed1 * 1 Items - 2.070,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Drain tap with tubing, ID 8 mmDrain tap with tubing, ID 8 mmJULA8970453PURA™ 10, PURA™ 14, PURA™ 22, PURA™ 30, SE-12, SE-26, SE-6, SE-Z, SL-12, SL-26, SL-6, SW22, SW23, HE-4, HL-4Log ind for tilgængelighed1 * 1 Items - 882,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Stainless steel hygiene insert for dentistry workStainless steel hygiene insert for dentistry workJULA9970532PURA™ 4Log ind for tilgængelighed1 * 1 Items - 850,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Stents lifter for dentistry workStents lifter for dentistry work461-1026PURA™ 4Log ind for tilgængelighed1 * 1 Items - 913,00 kr