Orders LinkContactUsLinkComponent
Water baths, PURA™
Water baths, PURA™
PURA 4, 18...99 °C, ±0,15 °C, 0,5 kW, 0,8 - 4,5 L, 120×270×140 mm, 210×350×220 mm
Catalog # 462-0625
Supplier:   JULABO GmbH
Water baths, PURA™
Catalog # 462-0625
Supplier:  JULABO GmbH
Water baths, PURA™
Water baths, PURA™
PURA 4, 18...99 °C, ±0,15 °C, 0,5 kW, 0,8 - 4,5 L, 120×270×140 mm, 210×350×220 mm
Price
7.170,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
PURA stainless steel water baths are functional and reliable for everyday operations due to their high material and component quality. The easy to use PURA water baths offer a number of functions and features that enable efficient use in the laboratory.
  • Intuitive, straightforward menu and bright, easy to read LED display
  • Splash-proof protected power switch
  • Integral drain screw (except PURA 4) and integrated timer
  • Built-in dry running protection
  • No interfering elements in the bath
  • Optical and acoustic alarm
  • Non slip rubber feet and ergonomic recessed grips
Units feature specially designed edge surfaces, which automatically drain condensed fluid back into the bath. Supplied with a removable platform which allows sample containers placed on the platform to be fully immersed and thus optimally tempered. PURA water baths are designed to enable natural circulation within the tank, all without a pump, guarantying homogeneous temperatures.
Leveringsinformation: Supplied with platform but without lids, a wide range of optional accessories are available. Note PURA™ 4, 10, 14, 22 and 30 baths accommodate a maximum of 1, 2, 3, 5 and 7 optional test tube racks respectively.
Documents
Specifications
  • Opvarmnings kapacitet:
    0,5 kW
  • Temperaturområde:
    18...99 °C
  • Kapacitet:
    0,8 - 4,5 L
  • Seq:
    001
  • B×D×H:
    210×350×220 mm
  • Bad B×D×H:
    120×270×140 mm
  • Temp. stabilitet:
    ±0,15 °C
  • Model:
    PURA 4
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970577
    Flat bath lid, ring sets with 3 openings, 190 mm Ø
    PURA™ 30
    Log in for availability
    1 * 1 Items - 2.350,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-3619
    Hule PP-kugler, 20 mm Ø
    Åbne varmebade, termostatbade, vandbade og rystevandbade
    Log in for availability
    1 * 1.000 Items - 1.090,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 462-0499
    Liquid level/cooling set
    PURA™ 10, PURA™ 14, PURA™ 22, PURA™30, SW22, SW23
    Log in for availability
    1 * 1 Items - 2.300,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970574
    Flat bath lid, ring sets with 6 openings, 115 mm Ø
    PURA™ 22
    Log in for availability
    1 * 1 Items - 2.840,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970575
    Flat bath lid, ring sets with 2 openings, 190 mm Ø
    PURA™ 22
    Log in for availability
    1 * 1 Items - 1.930,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970573
    Flat bath lid, ring sets with 8 openings, 92 mm Ø
    PURA™ 22
    Log in for availability
    1 * 1 Items - 3.390,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-1449
    Transparent lift up lid, for use up to +100 °C
    PURA™ 4
    Log in for availability
    1 * 1 Items - 1.270,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-1450
    Transparent lift up lid, for use up to +100 °C
    PURA™ 10
    Log in for availability
    1 * 1 Items - 1.590,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970534
    Cooling coil for working near ambient temperature
    PURA™ 10, PURA™ 14
    Log in for availability
    1 * 1 Items - 1.700,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970536
    Cooling coil for working near ambient temperature
    PURA™ 22, PURA™ 30
    Log in for availability
    1 * 1 Items - 2.010,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970570
    Flat bath lid, ring sets with 4 openings, 92 mm Ø
    PURA™ 10
    Log in for availability
    1 * 1 Items - 1.850,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-1453
    Transparent lift up lid, for use up to +100 °C
    PURA™ 30
    Log in for availability
    1 * 1 Items - 2.500,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-0943
    Plastic test tube rack for 21 tubes with 30 mm Ø
    B13, B17, B19, B27, B33, BT19, BT27, PURA™
    Log in for availability
    1 * 1 Items - 481,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-0942
    Plastic test tube rack for 90 tubes with 12/13 mm Ø, height 75 mm
    B13, B17, B19, B27, B33, BT19, BT27, PURA™
    Log in for availability
    1 * 1 Items - 481,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970571
    Flat bath lid, ring sets with 1 opening, 190 mm Ø
    PURA™ 10
    Log in for availability
    1 * 1 Items - 1.140,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-0941
    Plastic test tube rack for 60 tubes with 17 mm Ø, height 100 mm
    B13, B17, B19, B27, B33, BT19, BT27, PURA™
    Log in for availability
    1 * 1 Items - 481,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970576
    Flat bath lid, ring sets with 10 openings, 115 mm Ø
    PURA™ 30
    Log in for availability
    1 * 1 Items - 4.700,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-1451
    Transparent lift up lid, for use up to +100 °C
    PURA™ 14
    Log in for availability
    1 * 1 Items - 1.700,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970572
    Flat bath lid, ring sets with 6 openings, 92 mm Ø
    PURA™ 14
    Log in for availability
    1 * 1 Items - 2.630,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-1452
    Transparent lift up lid, for use up to +100 °C
    PURA™ 22
    Log in for availability
    1 * 1 Items - 2.070,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA8970453
    Drain tap with tubing, ID 8 mm
    PURA™ 10, PURA™ 14, PURA™ 22, PURA™ 30, SE-12, SE-26, SE-6, SE-Z, SL-12, SL-26, SL-6, SW22, SW23, HE-4, HL-4
    Log in for availability
    1 * 1 Items - 882,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • JULA9970532
    Stainless steel hygiene insert for dentistry work
    PURA™ 4
    Log in for availability
    1 * 1 Items - 850,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 461-1026
    Stents lifter for dentistry work
    PURA™ 4
    Log in for availability
    1 * 1 Items - 913,00 kr