n-Hexan ≥99%, AnalaR NORMAPUR® analytical reagent
Steel drum, 25 L
Leverandør: VWR Chemicals
n-Hexan ≥99%, AnalaR NORMAPUR® analytical reagent
Katalog # 24608.460
Leverandør: VWR Chemicals
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (1013 hPa)
- Smeltende Pt:–94,3 °C
- Tæthed:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-777-6
- FN:1208
- ADR:3,II
- NÅ:01-2119480412-44
Documents
Certifikater
Søg efter partinummer1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
Specifikationer
Specifikationstestresultater
- Assay:Min. 99.0 %
- Acidity:Max. 0.0004 meq/g
- Colouration:Max. 10 APHA
- Density (20/4):0.658 - 0.662
- Benzene:Max. 100 ppm
- Evaporation residue:Max. 10 ppm
- Thiophene:Max. 1 ppm
- Total S (as SO4):Max. 50 ppm
- Water:Max. 100 ppm
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.1 ppm
- Sn (Tin):Max. 0.1 ppm
- Zn (Zinc):Max. 0.1 ppm
Specifikationer
- Pakning:Steel drum
- Pakn:25 L



