Autoclavable goggles
Cleanroom goggles, Anti-UV, anti-fog, autoclavable, directly vented, Clear PC, blå
Leverandør: UNIVET
Gå til:
Product Details & Documents
The autoclavable goggles are designed for use in rooms with controlled contaminations or in aseptic conditions.
- Available with direct or indirect ventilation
- Elastic band in anti-static silicone with quick and smooth system
- RX insert
- Goggles with polycarbonate lens reusable up to 40 autoclave
The soft structure supports multiple cycles of steam, gamma or beta sterilisation without perceptible changes. The autoclavable goggles has a polycarbonate lens with anti-fog coating AF121 and anti-scratch.
Certifications: EN 166.
Leveringsinformation: Available with correction insert and replacement lens.
Documents
Specifikationer
- Model:Anti-UV, anti-fog, autoclavable, directly vented
- Farve:blå
- Beskrivelse:Cleanroom goggles
- Seq:03
- Linser:Clear PC
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Spare lens for 611S, clear PC
Produktinformation
- Katalog #TilgængelighedPrice
- Head band, silicone, 700×30×1,4 mm
Produktinformation
- Katalog #TilgængelighedPrice
- RX optical insert for goggles
Produktinformation
- Katalog #TilgængelighedPrice
- Spare lens, clear PC