Orders LinkContactUsLinkComponent
Ready-to-use tyramide amplification buffer, 1X
Ready-to-use tyramide amplification buffer, 1X
Ready-to-use tyramide amplification buffer, 1X, 25 mL
Katalog # 22027-25ML
Leverandør:   Biotium
Ready-to-use tyramide amplification buffer, 1X
Katalog # 22027-25ML
Leverandør:  Biotium
CAS nummer:  
Ready-to-use tyramide amplification buffer, 1X
Ready-to-use tyramide amplification buffer, 1X
Ready-to-use tyramide amplification buffer, 1X, 25 mL
CAS nummer:  
Price
1.310,00 kr
Log ind for tilgængelighed
1 * 25 mL
Product Details & Documents
For simple, optimised tyramide amplification of immunostaining in cells or tissue sections. No need to add hydrogen peroxide before use.
  • Ready-to-use
  • Simply dilute tyramide probe in buffer and proceed to signal development
  • Compatible with tyramides from Biotium or other suppliers
The Tyramide Amplification Buffer Plus (BTIU22029), a improved buffer for tyramide signal amplification provides significantly improved brightness, specificity, and sensitivity over this original buffer.
Simply mix the tyramide compound with the buffer to make a working solution for signal amplification. Although specifically optimised for our CF® Dye-, Biotin XX-, and DNP-tyramide products, Ready-to-Use Tyramide Amplification Buffer is also compatible with tyramide probes from other manufacturers.
Documents
Specifikationer
Specifikationer
  • Pakn:
    25 mL
  • Beskrivelse:
    Ready-to-use tyramide amplification buffer, 1X